Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCATTGGCCGAGGACTTGAGAGT[C/T]ACATTAGTAAGTTTGAAGCGCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 193245 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VDAC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VDAC2 - voltage dependent anion channel 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184783.2 | 684 | Missense Mutation | CAC,TAC | H,Y 24 | NP_001171712.1 | |
NM_001184823.1 | 684 | Intron | NP_001171752.1 | |||
NM_001324087.1 | 684 | Intron | NP_001311016.1 | |||
NM_001324088.1 | 684 | Intron | NP_001311017.1 | |||
NM_001324089.1 | 684 | Intron | NP_001311018.1 | |||
NM_001324090.1 | 684 | Intron | NP_001311019.1 | |||
NM_003375.4 | 684 | Intron | NP_003366.2 |