Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTCAGGAGACAGGGAGTCCCC[A/C]TCAAGAGAGGCTGCGGCTGACAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDPD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDPD5 - glycerophosphodiester phosphodiesterase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030792.6 | 3243 | UTR 3 | NP_110419.5 | |||
XM_006718697.3 | 3243 | UTR 3 | XP_006718760.1 | |||
XM_011545276.2 | 3243 | UTR 3 | XP_011543578.1 | |||
XM_011545277.2 | 3243 | UTR 3 | XP_011543579.1 | |||
XM_011545278.2 | 3243 | UTR 3 | XP_011543580.1 | |||
XM_011545279.2 | 3243 | UTR 3 | XP_011543581.1 | |||
XM_011545280.1 | 3243 | UTR 3 | XP_011543582.1 | |||
XM_011545281.2 | 3243 | UTR 3 | XP_011543583.1 | |||
XM_011545285.2 | 3243 | UTR 3 | XP_011543587.1 | |||
XM_011545286.1 | 3243 | UTR 3 | XP_011543588.1 | |||
XM_011545287.1 | 3243 | UTR 3 | XP_011543589.1 |
KLHL35 - kelch like family member 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039548.2 | 3243 | Intron | NP_001034637.2 | |||
XM_011544954.2 | 3243 | Intron | XP_011543256.1 |