Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAACTCCAGGCATTTCTGTTGG[A/G]GAGAAGGTCAGGGGAAAGGGTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616727 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF21B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF21B - PHD finger protein 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135862.2 | Intron | NP_001129334.1 | ||||
NM_001242450.1 | Intron | NP_001229379.1 | ||||
NM_001284296.1 | Intron | NP_001271225.1 | ||||
NM_138415.4 | Intron | NP_612424.1 | ||||
XM_006724122.1 | Intron | XP_006724185.1 | ||||
XM_011529855.1 | Intron | XP_011528157.1 |