Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCTCTCCCTCAGGCTCCAGCTT[A/G]GTGGCAGACTTCTTGTCAGAGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159350 MIM: 610709 | ||||||||||||||||||||
Literature Links: |
MCC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCC - mutated in colorectal cancers | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085377.1 | 1122 | Intron | NP_001078846.1 | |||
NM_002387.2 | 1122 | Intron | NP_002378.1 | |||
XM_005271991.2 | 1122 | Intron | XP_005272048.1 | |||
XM_017009473.1 | 1122 | Intron | XP_016864962.1 | |||
XM_017009474.1 | 1122 | Intron | XP_016864963.1 |
TSSK1B - testis specific serine kinase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032028.3 | 1122 | Silent Mutation | ACC,ACT | T,T 310 | NP_114417.1 |