Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTCTTTAGATATTTTTAGAA[A/G]GAAATATTAATGTCAATGATTTTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604068 | ||||||||||||||||||||
Literature Links: |
TPD52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPD52 - tumor protein D52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025252.2 | Intron | NP_001020423.1 | ||||
NM_001025253.2 | Intron | NP_001020424.1 | ||||
NM_001287140.1 | Intron | NP_001274069.1 | ||||
NM_001287142.1 | Intron | NP_001274071.1 | ||||
NM_001287143.1 | Intron | NP_001274072.1 | ||||
NM_001287144.1 | Intron | NP_001274073.1 | ||||
NM_005079.3 | Intron | NP_005070.1 |