Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAAGTCTCAGTGACATTTTCCA[A/G]ACATCTAACCTTGATTTGATTCTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601792 | |||||||||||||||||||||||
Literature Links: |
PPP1R2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PPP1R2 - protein phosphatase 1 regulatory inhibitor subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291504.1 | Intron | NP_001278433.1 | ||||
NM_001291505.1 | Intron | NP_001278434.1 | ||||
NM_001316325.1 | Intron | NP_001303254.1 | ||||
NM_006241.7 | Intron | NP_006232.1 | ||||
XM_006713682.3 | Intron | XP_006713745.1 |