Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGGTGCAACCGTCATGTGGAGA[G/T]CATCGACAAGCGCCACTGCTCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC1 - leucine rich repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018214.4 | 616 | Missense Mutation | AGC,ATC | S,I 16 | NP_060684.4 | |
XM_011514726.2 | 616 | Missense Mutation | AGC,ATC | S,I 16 | XP_011513028.1 | |
XM_011514727.2 | 616 | Intron | XP_011513029.1 | |||
XM_017010997.1 | 616 | Missense Mutation | AGC,ATC | S,I 16 | XP_016866486.1 |