Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGTGGCCCCAGGTGCCACCAAT[A/G]AAAATCCTACAGAAAATTCTCTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107710 MIM: 107741 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APOC1 - apolipoprotein C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321065.1 | 551 | UTR 3 | NP_001307994.1 | |||
NM_001321066.1 | 551 | UTR 3 | NP_001307995.1 | |||
NM_001645.4 | 551 | UTR 3 | NP_001636.1 |
APOC1P1 - apolipoprotein C1 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOE - apolipoprotein E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |