Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTTGTTTTAAAAATTATTTTCA[A/G]AGATATTCCACTTAAAGTTTCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607389 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SSBP2 - single stranded DNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256732.1 | Intron | NP_001243661.1 | ||||
NM_001256733.1 | Intron | NP_001243662.1 | ||||
NM_001256734.1 | Intron | NP_001243663.1 | ||||
NM_001256735.1 | Intron | NP_001243664.1 | ||||
NM_001256736.1 | Intron | NP_001243665.1 | ||||
NM_012446.3 | Intron | NP_036578.2 | ||||
XM_011543297.2 | Intron | XP_011541599.1 | ||||
XM_017009304.1 | Intron | XP_016864793.1 | ||||
XM_017009305.1 | Intron | XP_016864794.1 | ||||
XM_017009306.1 | Intron | XP_016864795.1 | ||||
XM_017009307.1 | Intron | XP_016864796.1 | ||||
XM_017009308.1 | Intron | XP_016864797.1 | ||||
XM_017009309.1 | Intron | XP_016864798.1 | ||||
XM_017009310.1 | Intron | XP_016864799.1 |