Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPNS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPNS3 - sphingolipid transporter 3 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320449.1 | Intron | NP_001307378.1 | ||||
NM_182538.4 | Intron | NP_872344.3 | ||||
XM_011523723.1 | Intron | XP_011522025.1 | ||||
XM_011523724.1 | Intron | XP_011522026.1 | ||||
XM_011523725.1 | Intron | XP_011522027.1 | ||||
XM_011523726.2 | Intron | XP_011522028.1 | ||||
XM_011523728.2 | Intron | XP_011522030.1 | ||||
XM_017024345.1 | Intron | XP_016879834.1 |