Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAAGGCGAGCTGTGATATATTTT[C/G]ATGGTGGTGGTTTTTGTTTTGGAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
AADACL2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AADACL2 - arylacetamide deacetylase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207365.3 | 451 | Missense Mutation | CAT,GAT | H,D 111 | NP_997248.2 | |
XM_011512753.1 | 451 | Missense Mutation | CAT,GAT | H,D 64 | XP_011511055.1 |