Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATAATTTTGGCAGGATTGTATG[A/G]TCTTTGAGGATGTGGCCATATATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF749 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF749 - zinc finger protein 749 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001023561.3 | 1279 | Missense Mutation | ATC,GTC | I,V 9 | NP_001018855.2 | |
NM_001321952.1 | 1279 | Missense Mutation | ATC,GTC | I,V 2 | NP_001308881.1 | |
NM_001321953.1 | 1279 | Intron | NP_001308882.1 | |||
NM_001321954.1 | 1279 | Intron | NP_001308883.1 | |||
XM_011526956.2 | 1279 | Missense Mutation | ATC,GTC | I,V 2 | XP_011525258.1 | |
XM_017026802.1 | 1279 | Missense Mutation | ATC,GTC | I,V 9 | XP_016882291.1 |