Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCACCTACCCTTGGAATCCTGGA[C/T]ATCATTGCACAGTACCCACTTCGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606934 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFAF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDUFAF1 - NADH:ubiquinone oxidoreductase complex assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016013.3 | 886 | Missense Mutation | ATA,ATG | I,M 186 | NP_057097.2 | |
XM_006720555.2 | 886 | Missense Mutation | ATA,ATG | I,M 186 | XP_006720618.1 | |
XM_011521658.1 | 886 | Missense Mutation | ATA,ATG | I,M 186 | XP_011519960.1 | |
XM_011521659.2 | 886 | Missense Mutation | ATA,ATG | I,M 186 | XP_011519961.1 | |
XM_017022293.1 | 886 | Intron | XP_016877782.1 |