Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATACTAAATGGTTTTTGCCAGTA[A/G]CAAAATTTTCATGTTTTATAGGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEUP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEUP1 - deuterosome assembly protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181645.3 | Intron | NP_857596.2 | ||||
XM_005273802.1 | Intron | XP_005273859.1 | ||||
XM_011542629.2 | Intron | XP_011540931.1 | ||||
XM_011542630.1 | Intron | XP_011540932.1 | ||||
XM_011542631.2 | Intron | XP_011540933.1 | ||||
XM_011542632.2 | Intron | XP_011540934.1 | ||||
XM_011542633.2 | Intron | XP_011540935.1 | ||||
XM_011542635.2 | Intron | XP_011540937.1 | ||||
XM_011542636.2 | Intron | XP_011540938.1 | ||||
XM_011542637.2 | Intron | XP_011540939.1 | ||||
XM_011542638.2 | Intron | XP_011540940.1 | ||||
XM_017017245.1 | Intron | XP_016872734.1 |
LOC102723746 - uncharacterized LOC102723746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |