Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAATCTTTGTTGATTTTAGTCT[C/T]CTGCTTTACTTAGAGTTGCACTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf84 - chromosome 9 open reading frame 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080551.2 | Intron | NP_001074020.2 | ||||
NM_173521.4 | Intron | NP_775792.4 | ||||
XM_006716974.2 | Intron | XP_006717037.1 | ||||
XM_006716975.2 | Intron | XP_006717038.1 | ||||
XM_011518302.2 | Intron | XP_011516604.1 | ||||
XM_011518303.1 | Intron | XP_011516605.1 | ||||
XM_011518306.1 | Intron | XP_011516608.1 | ||||
XM_011518309.1 | Intron | XP_011516611.1 | ||||
XM_017014340.1 | Intron | XP_016869829.1 | ||||
XM_017014341.1 | Intron | XP_016869830.1 |