Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACCACTTCCAGGCGGTCACGTC[C/T]GAGCTGCCCAGGCAGTCCTTCTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600856 MIM: 603240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN1C - cyclin dependent kinase inhibitor 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A18AS - solute carrier family 22 member 18 antisense | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302862.1 | 2538 | Missense Mutation | AGA,GGA | R,G 36 | NP_001289791.1 | |
NM_007105.3 | 2538 | Missense Mutation | AGA,GGA | R,G 139 | NP_009036.2 | |
XM_017017832.1 | 2538 | Missense Mutation | AGA,GGA | R,G 92 | XP_016873321.1 | |
XM_017017833.1 | 2538 | Missense Mutation | AGA,GGA | R,G 73 | XP_016873322.1 | |
XM_017017834.1 | 2538 | Missense Mutation | AGA,GGA | R,G 36 | XP_016873323.1 | |
XM_017017835.1 | 2538 | Missense Mutation | AGA,GGA | R,G 36 | XP_016873324.1 | |
XM_017017836.1 | 2538 | Missense Mutation | AGA,GGA | R,G 36 | XP_016873325.1 |