Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCGCCCATGGCGCAGCCACACA[A/C]GACCTCGGTCCCGTACTTCGCGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613814 MIM: 614535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC19 - tetratricopeptide repeat domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271420.1 | 123 | UTR 5 | NP_001258349.1 | |||
NM_017775.3 | 123 | UTR 5 | NP_060245.3 | |||
XM_017024801.1 | 123 | Missense Mutation | AAG,ACG | K,T 6 | XP_016880290.1 | |
XM_017024802.1 | 123 | Missense Mutation | AAG,ACG | K,T 6 | XP_016880291.1 |
ZSWIM7 - zinc finger SWIM-type containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042697.1 | 123 | Intron | NP_001036162.1 | |||
NM_001042698.1 | 123 | Intron | NP_001036163.1 |