Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGACTACCCTGCGGCGGCGCGG[C/T]TCGCAGTCCTTCTCAGCATGGACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609385 MIM: 142810 MIM: 600549 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DND1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DND1 - DND microRNA-mediated repression inhibitor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HARS - histidyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IK - IK cytokine, down-regulator of HLA II | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR55 - WD repeat domain 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017706.4 | 97 | UTR 5 | NP_060176.2 | |||
XM_005268469.2 | 97 | UTR 5 | XP_005268526.1 | |||
XM_017009600.1 | 97 | Intron | XP_016865089.1 |