Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACTCTAATTTTCTCTTACCAT[A/T]TGGGAAATTTCTGTTTCCTTGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615364 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK7 - adenylate kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152327.3 | Intron | NP_689540.2 | ||||
XM_006720021.2 | Intron | XP_006720084.1 | ||||
XM_006720022.2 | Intron | XP_006720085.1 | ||||
XM_011536403.2 | Intron | XP_011534705.1 | ||||
XM_017020956.1 | Intron | XP_016876445.1 | ||||
XM_017020957.1 | Intron | XP_016876446.1 | ||||
XM_017020958.1 | Intron | XP_016876447.1 | ||||
XM_017020959.1 | Intron | XP_016876448.1 | ||||
XM_017020960.1 | Intron | XP_016876449.1 |