Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGAGGGAGTGGGGGGAGCCCTG[A/T]GCTGCGCCCGGGAGGCTGGGGAGCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
LRRC3B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LRRC3B - leucine rich repeat containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317808.1 | 2 | Intron | NP_001304737.1 | |||
NM_001317809.1 | 2 | Intron | NP_001304738.1 | |||
NM_001317810.1 | 2 | Intron | NP_001304739.1 | |||
NM_001317811.1 | 2 | Intron | NP_001304740.1 | |||
NM_052953.3 | 2 | Intron | NP_443185.1 | |||
XM_005264849.3 | 2 | Intron | XP_005264906.1 | |||
XM_005264850.4 | 2 | Intron | XP_005264907.1 | |||
XM_011533332.2 | 2 | Intron | XP_011531634.1 | |||
XM_017005675.1 | 2 | UTR 5 | XP_016861164.1 |