Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACCTGACTTCTCAGAGTACAGG[A/G]AGCGGCCAGGTGTAGTCGCTCATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608801 MIM: 611487 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCDH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GCDH - glutaryl-CoA dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000159.3 | Intron | NP_000150.1 | ||||
NM_013976.3 | Intron | NP_039663.1 | ||||
XM_006722721.2 | Intron | XP_006722784.1 | ||||
XM_011527899.2 | Intron | XP_011526201.1 | ||||
XM_011527900.2 | Intron | XP_011526202.1 | ||||
XM_017026580.1 | Intron | XP_016882069.1 |
SYCE2 - synaptonemal complex central element protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105578.1 | Intron | NP_001099048.1 | ||||
XM_005259848.4 | Intron | XP_005259905.1 | ||||
XM_011527882.2 | Intron | XP_011526184.1 | ||||
XM_011527883.2 | Intron | XP_011526185.1 |
Set Membership: |
HapMap |