Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGTTGATGAAAGGCGTGTTGAC[C/T]GAGGCCTGCAATTCAAGGACAAGAA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 609169 MIM: 609969 | |||||||||||||||||||||||||||||
Literature Links: |
GAPDHS PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GAPDHS - glyceraldehyde-3-phosphate dehydrogenase, spermatogenic | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBSN - suprabasin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166034.1 | 1746 | Silent Mutation | NP_001159506.1 | |||
NM_001166035.1 | 1746 | Silent Mutation | NP_001159507.1 | |||
NM_198538.3 | 1746 | Silent Mutation | NP_940940.1 | |||
XM_011526929.2 | 1746 | Silent Mutation | XP_011525231.1 | |||
XM_011526930.2 | 1746 | Silent Mutation | XP_011525232.1 | |||
XM_011526931.2 | 1746 | Silent Mutation | XP_011525233.1 |