Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611143 MIM: 606769 MIM: 611983 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM175A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FAM175A - family with sequence similarity 175 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HELQ - helicase, POLQ-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297755.1 | Intron | NP_001284684.1 | ||||
NM_001297756.1 | Intron | NP_001284685.1 | ||||
NM_001297757.1 | Intron | NP_001284686.1 | ||||
NM_001297758.1 | Intron | NP_001284687.1 | ||||
NM_001297759.1 | Intron | NP_001284688.1 | ||||
NM_133636.3 | Intron | NP_598375.2 | ||||
XM_005262711.1 | Intron | XP_005262768.1 | ||||
XM_005262713.2 | Intron | XP_005262770.1 | ||||
XM_006714076.2 | Intron | XP_006714139.1 | ||||
XM_011531580.2 | Intron | XP_011529882.1 | ||||
XM_017007679.1 | Intron | XP_016863168.1 | ||||
XM_017007680.1 | Intron | XP_016863169.1 | ||||
XM_017007681.1 | Intron | XP_016863170.1 | ||||
XM_017007682.1 | Intron | XP_016863171.1 | ||||
XM_017007683.1 | Intron | XP_016863172.1 |
MRPS18C - mitochondrial ribosomal protein S18C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297767.1 | Intron | NP_001284696.1 | ||||
NM_001297769.1 | Intron | NP_001284698.1 | ||||
NM_001297770.1 | Intron | NP_001284699.1 | ||||
NM_016067.3 | Intron | NP_057151.1 |
Set Membership: |
HapMap Validated |