Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604366 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAI1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DNAI1 - dynein axonemal intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281428.1 | 77 | UTR 5 | NP_001268357.1 | |||
NM_012144.3 | 77 | UTR 5 | NP_036276.1 | |||
XM_006716758.3 | 77 | Intron | XP_006716821.1 | |||
XM_011517846.1 | 77 | UTR 5 | XP_011516148.1 | |||
XM_011517847.2 | 77 | UTR 5 | XP_011516149.1 | |||
XM_011517848.1 | 77 | UTR 5 | XP_011516150.1 | |||
XM_011517849.1 | 77 | UTR 5 | XP_011516151.1 | |||
XM_011517850.2 | 77 | UTR 5 | XP_011516152.1 | |||
XM_017014625.1 | 77 | UTR 5 | XP_016870114.1 |
FAM219A - family with sequence similarity 219 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184940.1 | 77 | Intron | NP_001171869.1 | |||
NM_001184941.1 | 77 | Intron | NP_001171870.1 | |||
NM_001184942.1 | 77 | Intron | NP_001171871.1 | |||
NM_001184943.1 | 77 | Intron | NP_001171872.1 | |||
NM_001184945.1 | 77 | Intron | NP_001171874.1 | |||
NM_147202.1 | 77 | Intron | NP_671735.1 | |||
XM_005251402.4 | 77 | Intron | XP_005251459.1 | |||
XM_011517797.2 | 77 | Intron | XP_011516099.1 | |||
XM_011517798.2 | 77 | Intron | XP_011516100.1 | |||
XM_011517799.2 | 77 | Intron | XP_011516101.1 |
Set Membership: |
HapMap Validated |