Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611479 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC121 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCDC121 - coiled-coil domain containing 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142683.2 | 56 | Intron | NP_001136155.1 | |||
NM_024584.4 | 56 | Intron | NP_078860.2 | |||
XM_005264560.3 | 56 | Intron | XP_005264617.1 |
GPN1 - GPN-loop GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145047.1 | 56 | Intron | NP_001138519.1 | |||
NM_001145048.1 | 56 | Intron | NP_001138520.1 | |||
NM_001145049.1 | 56 | Intron | NP_001138521.1 | |||
NM_007266.3 | 56 | Missense Mutation | AAG,AGG | K,R 12 | NP_009197.2 |
ZNF512 - zinc finger protein 512 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |