Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609479 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZAK PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZAK - sterile alpha motif and leucine zipper containing kinase AZK | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016653.2 | Intron | NP_057737.2 | ||||
NM_133646.2 | Intron | NP_598407.1 | ||||
XM_005246640.2 | Intron | XP_005246697.1 | ||||
XM_017004323.1 | Intron | XP_016859812.1 | ||||
XM_017004324.1 | Intron | XP_016859813.1 |
Set Membership: |
HapMap Validated |