Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605251 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ABCC5 - ATP binding cassette subfamily C member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001023587.2 | 5981 | Intron | NP_001018881.1 | |||
NM_001320032.1 | 5981 | UTR 3 | NP_001306961.1 | |||
NM_005688.3 | 5981 | UTR 3 | NP_005679.2 | |||
XM_005247058.4 | 5981 | UTR 3 | XP_005247115.1 | |||
XM_005247059.4 | 5981 | UTR 3 | XP_005247116.1 | |||
XM_011512314.1 | 5981 | UTR 3 | XP_011510616.1 | |||
XM_011512315.1 | 5981 | Intron | XP_011510617.1 | |||
XM_017005492.1 | 5981 | UTR 3 | XP_016860981.1 | |||
XM_017005493.1 | 5981 | Intron | XP_016860982.1 | |||
XM_017005494.1 | 5981 | Intron | XP_016860983.1 |
Set Membership: |
HapMap Validated |