Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTTATGTATATAAGGTGAGCGT[A/G]CATTTATGTACATTTCCAGTAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF38B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRPF38B - pre-mRNA processing factor 38B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018061.2 | Intron | NP_060531.2 | ||||
XM_011541687.2 | Intron | XP_011539989.1 | ||||
XM_017001596.1 | Intron | XP_016857085.1 | ||||
XM_017001597.1 | Intron | XP_016857086.1 | ||||
XM_017001598.1 | Intron | XP_016857087.1 |