Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGCCGCCATCTTGGCAAGAGG[C/T]GAAGCGGCAGCGGTTCCTGTCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARCN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARCN1 - archain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142281.1 | 37 | UTR 5 | NP_001135753.1 | |||
NM_001655.4 | 37 | UTR 5 | NP_001646.2 | |||
XM_005271542.3 | 37 | UTR 5 | XP_005271599.1 |
IFT46 - intraflagellar transport 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168618.1 | 37 | Intron | NP_001162089.1 | |||
NM_020153.3 | 37 | Intron | NP_064538.3 | |||
XM_011542905.2 | 37 | Intron | XP_011541207.1 | |||
XM_011542906.2 | 37 | Intron | XP_011541208.1 | |||
XM_017018017.1 | 37 | Intron | XP_016873506.1 | |||
XM_017018018.1 | 37 | Intron | XP_016873507.1 |