Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607129 MIM: 603498 | ||||||||||||||||||||
Literature Links: |
MICAL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MICAL1 - microtubule associated monooxygenase, calponin and LIM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159291.1 | Intron | NP_001152763.1 | ||||
NM_001286613.1 | Intron | NP_001273542.1 | ||||
NM_022765.3 | Intron | NP_073602.3 |
PPIL6 - peptidylprolyl isomerase like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMPD2 - sphingomyelin phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003080.2 | Intron | NP_003071.2 |