Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCCACAGCCCAACGGAGGTGG[C/G]CGGGACCTCAGCCTCCAGTCCCGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604295 | ||||||||||||||||||||
Literature Links: |
VAX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VAX2 - ventral anterior homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012476.2 | 211 | Missense Mutation | GCC,GGC | A,G 47 | NP_036608.1 | |
XM_006711982.3 | 211 | Missense Mutation | GCC,GGC | A,G 47 | XP_006712045.1 | |
XM_011532750.2 | 211 | Missense Mutation | GCC,GGC | A,G 47 | XP_011531052.1 | |
XM_011532751.2 | 211 | Missense Mutation | GCC,GGC | A,G 47 | XP_011531053.1 |