Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATAATCGCCACTAGCTGTGGAG[C/T]TGCTGCCGTAGGCTGGGCACCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604551 MIM: 613497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CH25H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CH25H - cholesterol 25-hydroxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIPA - lipase A, lysosomal acid type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000235.3 | Intron | NP_000226.2 | ||||
NM_001127605.2 | Intron | NP_001121077.1 | ||||
NM_001288979.1 | Intron | NP_001275908.1 |