Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGAAGCACAAAACAGTGAGACA[A/G]CCATCCTTCTATTTTCAATTATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALCRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALCRL - calcitonin receptor like receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271751.1 | 1865 | UTR 3 | NP_001258680.1 | |||
NM_005795.5 | 1865 | UTR 3 | NP_005786.1 | |||
XM_005246231.3 | 1865 | UTR 3 | XP_005246288.2 | |||
XM_005246232.3 | 1865 | UTR 3 | XP_005246289.2 | |||
XM_005246234.3 | 1865 | UTR 3 | XP_005246291.2 | |||
XM_017003151.1 | 1865 | UTR 3 | XP_016858640.1 |