Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGACCTCCCCGCAGGGAGTGCGG[A/G]GCGGCAGTCGCGGAAGGGACAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCTP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCTP - phosphatidylcholine transfer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102402.2 | 188 | UTR 5 | NP_001095872.1 | |||
NM_021213.3 | 188 | Intron | NP_067036.2 | |||
XM_005257549.1 | 188 | Intron | XP_005257606.1 | |||
XM_005257550.3 | 188 | Intron | XP_005257607.1 | |||
XM_017024913.1 | 188 | UTR 5 | XP_016880402.1 |