Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606288 MIM: 606289 MIM: 606290 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDHGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCDHGA1 - protocadherin gamma subfamily A, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018912.2 | 764 | Intron | NP_061735.1 | |||
NM_031993.1 | 764 | Intron | NP_114382.1 |
PCDHGA2 - protocadherin gamma subfamily A, 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018915.3 | 764 | Silent Mutation | AGA,AGG | R,R 184 | NP_061738.1 | |
NM_032009.2 | 764 | Silent Mutation | AGA,AGG | R,R 184 | NP_114398.1 |
PCDHGA3 - protocadherin gamma subfamily A, 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |