Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGTTCTGTGCCACAGCCTAGC[A/G]GAGGGGAAAGAAAGGGAGAGTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300023 MIM: 300538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP4 - Rho GTPase activating protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164741.1 | Intron | NP_001158213.1 | ||||
NM_001666.4 | Intron | NP_001657.3 |
AVPR2 - arginine vasopressin receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000054.4 | Intron | NP_000045.1 | ||||
NM_001146151.1 | Intron | NP_001139623.1 | ||||
XM_006724828.3 | Intron | XP_006724891.1 |