Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATAATGTGGTGTGGACTATCA[A/G]TTTATTTTATATTTTCATGTTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSFA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SSFA2 - sperm specific antigen 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130445.2 | Intron | NP_001123917.1 | ||||
NM_001287503.1 | Intron | NP_001274432.1 | ||||
NM_001287504.1 | Intron | NP_001274433.1 | ||||
NM_001287505.1 | Intron | NP_001274434.1 | ||||
NM_006751.6 | Intron | NP_006742.2 | ||||
XM_005246812.1 | Intron | XP_005246869.1 | ||||
XM_005246813.1 | Intron | XP_005246870.1 | ||||
XM_011511701.2 | Intron | XP_011510003.1 | ||||
XM_011511702.2 | Intron | XP_011510004.1 | ||||
XM_011511703.2 | Intron | XP_011510005.1 | ||||
XM_017004782.1 | Intron | XP_016860271.1 |