Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAGACGGCAATCGACCAGACGG[C/G]GATCGGGAGCTACTACCAGCTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD53 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD53 - ankyrin repeat domain 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115116.1 | 351 | Missense Mutation | GCG,GGG | A,G 105 | NP_001108588.1 | |
NM_024933.3 | 351 | Missense Mutation | GCG,GGG | A,G 105 | NP_079209.3 | |
XM_005264569.4 | 351 | Missense Mutation | GCG,GGG | A,G 71 | XP_005264626.1 | |
XM_011533114.2 | 351 | Missense Mutation | GCG,GGG | A,G 105 | XP_011531416.1 |
TEX261 - testis expressed 261 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |