Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTAGCAGCAGAGCCAGGGAAAG[C/T]GGCGCACTCTTCAGTTACAGCCAGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607703 | |||||||||||||||||||||||
Literature Links: |
NUP210 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NUP210 - nucleoporin 210 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024923.3 | Intron | NP_079199.2 | ||||
XM_005264994.3 | Intron | XP_005265051.1 | ||||
XM_011533538.2 | Intron | XP_011531840.1 | ||||
XM_017006018.1 | Intron | XP_016861507.1 | ||||
XM_017006019.1 | Intron | XP_016861508.1 | ||||
XM_017006020.1 | Intron | XP_016861509.1 | ||||
XM_017006021.1 | Intron | XP_016861510.1 |