Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGTCTTCGTCCGGAATATTGAA[C/T]ACTTGAAGAACGGCAGCCTAATATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612903 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCN9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LCN9 - lipocalin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001676.1 | 193 | Missense Mutation | CAC,TAC | H,Y 65 | NP_001001676.1 | |
XM_017014712.1 | 193 | Missense Mutation | CAC,TAC | H,Y 65 | XP_016870201.1 |
Set Membership: |
HapMap |