Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCGTTGCAGAGGTAAATGGTTTT[C/G]AGTAGCAGTTATATTGAATACTGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604496 MIM: 604174 | |||||||||||||||||||||||
Literature Links: |
NKIRAS1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NKIRAS1 - NFKB inhibitor interacting Ras like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL15 - ribosomal protein L15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253379.1 | Intron | NP_001240308.1 | ||||
NM_001253380.1 | Intron | NP_001240309.1 | ||||
NM_001253382.1 | Intron | NP_001240311.1 | ||||
NM_001253383.1 | Intron | NP_001240312.1 | ||||
NM_001253384.1 | Intron | NP_001240313.1 | ||||
NM_002948.3 | Intron | NP_002939.2 |