Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCATGTCACAGCACTTTGCATT[C/T]ATGATTATTAGCTTGAAACAGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607496 MIM: 609395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOSTRIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOSTRIN - nitric oxide synthase trafficking | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039724.3 | 2593 | UTR 3 | NP_001034813.2 | |||
NM_001171631.1 | 2593 | UTR 3 | NP_001165102.1 | |||
NM_001171632.1 | 2593 | UTR 3 | NP_001165103.1 | |||
NM_052946.3 | 2593 | UTR 3 | NP_443178.2 | |||
XM_005246270.1 | 2593 | Intron | XP_005246327.1 | |||
XM_006712233.3 | 2593 | Intron | XP_006712296.1 | |||
XM_011510540.2 | 2593 | Intron | XP_011508842.1 | |||
XM_011510543.2 | 2593 | Intron | XP_011508845.1 | |||
XM_017003278.1 | 2593 | Intron | XP_016858767.1 | |||
XM_017003279.1 | 2593 | Intron | XP_016858768.1 | |||
XM_017003280.1 | 2593 | Intron | XP_016858769.1 |
SPC25 - SPC25, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020675.3 | 2593 | Intron | NP_065726.1 | |||
XM_011511516.2 | 2593 | UTR 3 | XP_011509818.1 |