Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTGTTCTCTGGTCCTAGCTCCT[A/T]CCTATATGGTGGCAGAAGTTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154235 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAK - male germ cell associated kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM14B - transmembrane protein 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127711.2 | Intron | NP_001121183.1 | ||||
NM_001286484.1 | Intron | NP_001273413.1 | ||||
NM_001286488.1 | Intron | NP_001273417.1 | ||||
NM_001286489.1 | Intron | NP_001273418.1 | ||||
NM_030969.4 | Intron | NP_112231.3 |