Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300897 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC4H2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZC4H2 - zinc finger C4H2-type containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178032.2 | 588 | Silent Mutation | ACA,ACG | T,T 121 | NP_001171503.1 | |
NM_001178033.2 | 588 | Intron | NP_001171504.1 | |||
NM_001243804.1 | 588 | Silent Mutation | ACA,ACG | T,T 121 | NP_001230733.1 | |
NM_018684.3 | 588 | Silent Mutation | ACA,ACG | T,T 144 | NP_061154.1 |