Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604958 MIM: 611852 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTL6A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACTL6A - actin like 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004301.4 | 623 | Intron | NP_004292.1 | |||
NM_177989.3 | 623 | Intron | NP_817126.1 | |||
NM_178042.3 | 623 | Intron | NP_829888.1 |
MRPL47 - mitochondrial ribosomal protein L47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020409.2 | 623 | Silent Mutation | CTC,CTT | L,L 249 | NP_065142.2 | |
NM_177988.1 | 623 | Silent Mutation | CTC,CTT | L,L 139 | NP_817125.1 |
Set Membership: |
HapMap |