Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAACAAGCAAATCCTAGTGCT[A/G]GGCCTGGATGGAGCAGGAAAAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612405 MIM: 602122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL9 - ADP ribosylation factor like GTPase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206919.1 | 328 | UTR 5 | NP_996802.1 | |||
XM_005265719.3 | 328 | Silent Mutation | CTA,CTG | L,L 102 | XP_005265776.1 | |
XM_005265720.3 | 328 | UTR 5 | XP_005265777.1 | |||
XM_006714036.3 | 328 | UTR 5 | XP_006714099.1 |
SRP72 - signal recognition particle 72 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |