Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAAATTTTAAATGCTGAATAT[G/T]ATAAAAATCCCTCAAATCAGTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD8 - major facilitator superfamily domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152778.2 | 1433 | Intron | NP_689991.1 | |||
XM_005262893.1 | 1433 | Intron | XP_005262950.1 | |||
XM_005262896.1 | 1433 | Intron | XP_005262953.1 | |||
XM_005262897.2 | 1433 | Intron | XP_005262954.1 | |||
XM_005262898.2 | 1433 | UTR 3 | XP_005262955.1 | |||
XM_011531830.1 | 1433 | Intron | XP_011530132.1 | |||
XM_011531831.1 | 1433 | Intron | XP_011530133.1 | |||
XM_011531832.1 | 1433 | UTR 3 | XP_011530134.1 | |||
XM_017007989.1 | 1433 | UTR 3 | XP_016863478.1 |