Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGACTAAAATCTGATGATACATA[C/T]ATATATTTTTAATTTTGTCTTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147485 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPBP1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPBP1L1 - GC-rich promoter binding protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IPP - intracisternal A particle-promoted polypeptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM69 - transmembrane protein 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016486.3 | Intron | NP_057570.2 |