Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGGTCACCCTCTCTTCCCCAC[A/G]GCACTCCTCAGGCCCCTTCACTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606351 MIM: 611101 MIM: 603366 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESPN - espin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHG5 - pleckstrin homology and RhoGEF domain containing G5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042663.1 | Intron | NP_001036128.1 | ||||
NM_001042664.1 | Intron | NP_001036129.1 | ||||
NM_001042665.1 | Intron | NP_001036130.1 | ||||
NM_001265592.1 | Intron | NP_001252521.1 | ||||
NM_001265593.1 | Intron | NP_001252522.1 | ||||
NM_001265594.1 | Intron | NP_001252523.1 | ||||
NM_020631.4 | Intron | NP_065682.2 | ||||
NM_198681.3 | Intron | NP_941374.2 |
TNFRSF25 - TNF receptor superfamily member 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |