Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAAGCCCTCTGAAAGTTACTCC[C/T]TTACAACCTCATCATGAAGGACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf72 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C7orf72 - chromosome 7 open reading frame 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161834.2 | 807 | Silent Mutation | CCC,CCT | P,P 194 | NP_001155306.2 | |
XM_011515052.1 | 807 | Silent Mutation | CCC,CCT | P,P 194 | XP_011513354.1 | |
XM_011515053.2 | 807 | Silent Mutation | CCC,CCT | P,P 194 | XP_011513355.1 |